Skip to Content
Merck
All Photos(2)

Key Documents

375187

Sigma-Aldrich

Trimethyltin chloride solution

1.0 M in hexanes

Synonym(s):

Chlorotrimethylstannane, Chlorotrimethyltin, Trimethylchlorostannane, Trimethylchlorotin, Trimethylstannyl chloride, Trimethyltin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnCl
CAS Number:
Molecular Weight:
199.27
Beilstein:
3535111
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in hexanes

density

0.797 g/mL at 25 °C

SMILES string

C[Sn](C)(C)Cl

InChI

1S/3CH3.ClH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

KWTSZCJMWHGPOS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyltin chloride solution (1.0 M in hexanes) can be used as a reagent in the synthesis of conjugated polymers based on isoindigo as acceptor and syn- or anti-benzo[1,2-b:5,4-b′]difuran (BDF) as a donor by Stille cross-coupling reaction. It can further be used for the investigation of optical, electrochemical and photovoltaic properties.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photovoltaic properties of new conjugated polymers based on syn-and anti-benzodifuran
Hu C, et al.
Polym. Chem., 3(10), 2949-2955 (2012)
Xiaojiang Tang et al.
Toxicology, 271(1-2), 45-50 (2010-03-10)
Trimethyltin chloride (TMT), a byproduct of plastic stabilizers, has caused 67 poisoning accidents in the world; more than 98% (1814/1849) of the affected patients since 1998 have been in China. As a long-established toxic chemical, TMT severely affects the limbic
Kiyokazu Ogita et al.
Journal of neuroscience research, 82(5), 609-621 (2005-11-08)
The hippocampal dentate gyrus in adult animals is known to contain neural progenitors that proliferate and differentiate into neurons in response to brain injury. Little has been observed, however, on regeneration of the granule cell layer of the dentate gyrus
Katharina Krüger et al.
British journal of pharmacology, 144(2), 283-292 (2005-01-19)
1. Organotin compounds such as trimethyltin chloride (TMT) are among the most toxic of the organometallics. As their main target for toxicity is the central nervous system, the aim of the present study was to investigate the effects of TMT
Koichi Kawada et al.
Journal of neuroscience research, 86(7), 1635-1646 (2008-01-10)
Our earlier study demonstrated that in vivo acute treatment with trimethyltin chloride (TMT) produces severe neuronal damage in the dentate gyrus and cognition impairment in mice. In the present study, we assessed whether TMT was capable of causing neuronal degeneration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service