Skip to Content
Merck
All Photos(4)

Key Documents

372838

Sigma-Aldrich

Poly(ethylene oxide)

average MV 8,000,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene oxide), average Mv ~8,000,000 (nominal), powder

form

powder

mol wt

average Mv ~8,000,000 (nominal)

contains

200-500 ppm BHT as inhibitor

viscosity

10,000-15,000 cP, 1 % in H2O(25 °C)

functional group

hydroxyl

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Poly (ethylene oxide)-based electrolyte for solid-state lithium batteries with high voltage positive electrodes: This study evaluates the role of electrolyte oxidation in battery performance, highlighting the challenges and potential improvements for PEO-based solid polymer electrolytes (Homann et al., 2020).

  • Lithium bis (fluorosulfonyl) imide/poly (ethylene oxide) polymer electrolyte for all solid-state Li-S cell: Discusses the development of PEO-based polymer electrolytes for lithium-sulfur batteries, highlighting advances in energy storage technologies (Judez et al., 2017).

  • Poly (ethylene oxide)-based electrolytes for lithium-ion batteries: Explores PEO-based materials as candidates for solid-state electrolytes in lithium-ion batteries, addressing challenges in electrical vehicle and portable electronics industries ( He, Xie, 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D D Smyth et al.
Cardiovascular drugs and therapy, 4(1), 297-300 (1990-02-01)
Previous studies have demonstrated that Separan AP-30, a drag-reducing polymer, significantly decreased the formation of atherosclerotic plaques in rabbits fed a high-cholesterol diet. Furthermore, Separan AP-273, a polymer similar to but longer than Separan AP-30, markedly increased cardiac output in
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
P I Polimeni et al.
Journal of cardiovascular pharmacology, 14(3), 374-380 (1989-09-01)
The acute hemodynamic effects of an intravenously (i.v.) injected poly(ethylene oxide), Polyox WSR N-60K (dose 50 mg/kg), were studied in the open-chest rat anesthetized with sodium pentobarbital. The injectate is one of four drag-reducing polymers known to augment in vitro
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service