Skip to Content
Merck
All Photos(3)

Documents

369713

Sigma-Aldrich

1-Benzyl-2-methylimidazole

technical grade, 90%

Synonym(s):

1-Benzyl-2-methyl-1H-imidazole, 1-Phenylmethyl-2-methylimidazole, 2-Methyl-1-(phenylmethyl)-1H-imidazole, 2-Methyl-N-benzylimidazole, N-Benzyl-2-methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2
CAS Number:
Molecular Weight:
172.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

refractive index

n20/D 1.5645 (lit.)

bp

125-127 °C/3 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1nccn1Cc2ccccc2

InChI

1S/C11H12N2/c1-10-12-7-8-13(10)9-11-5-3-2-4-6-11/h2-8H,9H2,1H3

InChI key

FBHPRUXJQNWTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Benzyl-2-methylimidazole is a nitrogen heterocycle and forms mixed ligand Pt(II) complexes. 1-Benzyl-2-methylimidazole on treatement with ammonia and sodium yields 2-methylimidazole. Deposition of plasma-polymerized 1-benzyl-2-methylimidazole (PPBMI) thin films onto glass substrates by glow discharge technique have been reported.

Application

1-Benzyl-2-methylimidazole may be employed as catalyst to investigate the effect of catalysts on the curing reaction of the biphenyl epoxy (4,4′-diglycidyloxy-3,3′,5,5′-tetramethyl biphenyl)dicyclopentadiene type phenolic resin system.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
Han S, et al.
Journal of Applied Polymer Science, 68(7), 1125-1137 (1998)
Studies on imidazole compounds; a synthesis of imidazoles with functional groups in the 2-position.
R G JONES
Journal of the American Chemical Society, 71(2), 383-386 (1949-02-01)
Structural and Optical Properties of Plasma-Polymerized 1-BENZYL-2-METHYLIMIDAZOLE Thin Films.
Sarker RB and Bhuiyan AH.
International Journal of Modern Physics B, 25(14), 1941-1955 (2011)
Julien Sarju et al.
Dalton transactions (Cambridge, England : 2003), (39)(39), 5302-5312 (2008-10-02)
The nitrile ligands in trans-[PtX2(PhCN)2] (X = Cl, Br, I) undergo sequential 1,3 dipolar cycloadditions with nitrones R1R2C=N+(Me)-O(-) (R1 = H, R2 = Ph; R1 = CO2Et, R2 = CH2CO2Et) to selectively form the Delta4-1,2,4-oxadiazoline complexes trans-[PtX2(PhCN) (N=C(Ph)-O-N(Me)-CR1R2)] or trans-[PtX2(N=C(Ph)-O-N(Me)-CR1R2)2]
Rahul A Patil et al.
Analytical and bioanalytical chemistry, 410(19), 4645-4655 (2018-01-31)
The rapid expansion in the study and use of ionic liquids (ILs) is a result of their unique properties including negligible volatility, high thermal stability, and ability to dissolve disparate compounds. However, because ILs have infinitely variable structures (often referred

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service