Skip to Content
Merck
All Photos(1)

Key Documents

365203

Sigma-Aldrich

Lithium p-toluenesulfinate

98%

Synonym(s):

p-Toluenesulfinic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2Li
CAS Number:
Molecular Weight:
162.14
Beilstein:
4163977
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

impurities

≤2% Li2CO3

SMILES string

[Li+].Cc1ccc(cc1)S([O-])=O

InChI

1S/C7H8O2S.Li/c1-6-2-4-7(5-3-6)10(8)9;/h2-5H,1H3,(H,8,9);/q;+1/p-1

InChI key

MSUZXYWQEDRGFN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium p-toluenesulfinate may be used in the preparation of 2-oxo-1,3-diphenyl-1,2-dihydroquinoline-4-carbonitrile.[1] It may be used in the preparation of highly fluorescent compound, 3-amino-6-methoxy-4-p-tolylsulfonylquinolone.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Cyano-6, 7-dimethoxycarbostyrils with Solvent-and pH-Independent High Fluorescence Quantum Yields and Emission Maxima.
Ahvale AB, et al.
European Journal of Organic Chemistry, 3, 563-571 (2008)
Month 2013 6-Methoxy-2-oxo-1, 2-dihydroquinoline-3, 4-dicarbonitriles, A Red Compound Class with Solvent and pH Independent Green Fluorescence Maxima.
Enoua GC, et al.
Journal of Heterocyclic Chemistry, 51, 492-501 (2014)
Xianghong Li et al.
The journal of adhesive dentistry, 14(2), 129-136 (2011-09-22)
The use of sodium hypochlorite (NaOCl) can decrease the bond strength of 4-META/MMA-TBB resin cement to root dentin. The aim of this in vitro study was to evaluate the effect of sodium toluene sulfinic acid (SA) as a pretreatment to
K Eulitz et al.
Lipids, 34(8), 873-877 (1999-10-21)
Conjugated linoleic acid (CLA) mixtures were isomerized with p-toluenesulfinic acid or I2 catalyst. The resultant mixtures of the eight cis/trans geometric isomers of 8,10-, 9,11-, 10,12-, and 11,13-octadecadienoic (18:2) acid methyl esters were separated by silver ion-high-performance liquid chromatography (Ag+-HPLC)
Xiaoxiang Zhang et al.
The Journal of organic chemistry, 75(18), 6290-6293 (2010-08-28)
A general and efficient method to prepare 2,4-di- and trisubstituted thiazoles via p-TsOH·H(2)O-catalyzed cyclization of trisubstituted propargylic alcohols with thioamides is described. The reaction was accomplished in moderate to excellent product yields under mild conditions that did not require the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service