Skip to Content
Merck
All Photos(2)

Key Documents

360074

Sigma-Aldrich

Magnesium bromide

98%

Synonym(s):

Ddibromomagnesium, Magnesium bromide salt, Magnesium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgBr2
CAS Number:
Molecular Weight:
184.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
form:
powder

Quality Level

Assay

98%

form

powder

mp

711 °C (lit.)

density

3.72 g/mL at 25 °C (lit.)

SMILES string

Br[Mg]Br

InChI

1S/2BrH.Mg/h2*1H;/q;;+2/p-2

InChI key

OTCKOJUMXQWKQG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium bromide (MgBr2) can be used as a catalyst in:
  • Acetylation and benzoylation of alcohols (primary and secondary) with acid anhydrides.
  • In the synthesis of dihydropyrimidinones , trifluoromethylnaphthalenes , substituted 1,2,3,4 tetrahydropyrimidin-2-ones .
  • In the preparation of Aldol derivatives by hydrogenation of Baylis-Hillman olefins.
  • Deprotection of SEM (2-(Trimethylsilyl)ethoxymethyl) ether group.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of trifluoromethylnaphthalenes
Mellor JM, et al.
Tetrahedron, 56(51), 10067-10074 (2000)
Magnesium bromide mediated highly diastereoselective heterogeneous hydrogenation of olefins
Bouzide, Abderrahim
Organic Letters, 4(8), 1347-1350 (2002)
Efficient Magnesium Bromide-Catalyzed One-pot Synthesis of Substituted 1, 2, 3, 4-Tetrahydropyrimidin-2-ones Under Solvent-free Conditions
Salehi H and Guo Q-X
Chin. J. Chem., 23(1), 91-97 (2005)
A facile and efficient one-pot synthesis of dihydropyrimidinones catalyzed by magnesium bromide under solvent-free conditions
Salehi H and Guo Q-X
Synthetic Communications, 34(1), 171-179 (2004)
Magnesium bromide catalysed acylation of alcohols
Pansare SV, et al.
Synthetic Communications, 30(14), 2587-2592 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service