Skip to Content
Merck
All Photos(2)

Key Documents

359807

Sigma-Aldrich

Z-Phe-OH

99%

Synonym(s):

N-(Carbobenzyloxy)-L-phenylalanine, Z-L-Phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
299.32
Beilstein:
2222826
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D +5°, c = 5 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

85-87 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2

InChI

1S/C17H17NO4/c19-16(20)15(11-13-7-3-1-4-8-13)18-17(21)22-12-14-9-5-2-6-10-14/h1-10,15H,11-12H2,(H,18,21)(H,19,20)/t15-/m0/s1

InChI key

RRONHWAVOYADJL-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of thermolysin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhruvkumar Soni et al.
Biomaterials, 222, 119441-119441 (2019-09-01)
While antiretroviral therapy (ART) has revolutionized treatment and prevention of human immunodeficiency virus type one (HIV-1) infection, regimen adherence, viral mutations, drug toxicities and access stigma and fatigue are treatment limitations. These have led to new opportunities for the development
B Harris et al.
Biochimica et biophysica acta, 889(1), 1-5 (1986-10-31)
The possible role of metalloendoproteinase in stimulus-secretion coupling in adrenal chromaffin cells was examined using the metalloendoproteinase inhibitors 1,10-phenanthroline and carbobenzoxy-Gly-Phe-NH2. Catecholamine release elicited by nicotine or by depolarisation with 55 mM K+ was almost completely abolished by 0.5 mM
J Wiese et al.
Biomedica biochimica acta, 50(10-11), S90-S93 (1991-01-01)
The alpha-glyceryl esters of Z-Gly, Z-Phe and Z-Tyr were synthesized and their use for protease catalyzed peptide synthesis was studied. Three enzymes isolated from crude papain were compared in their catalytic potency. Syntheses with alpha-chymotrypsin were performed in a biphasic
S Stoeva
International journal of peptide and protein research, 37(4), 325-330 (1991-04-01)
Diethylpyrocarbonate (DEPC) inactivated the neutral zinc proteinase from Bacillus mesentericus strain 76/Bacillus subtilis (MCP 76) by ethoxycarbonylation completely. Exposure of the enzyme to DEPC together with the competitive inhibitor Z-L-phenylalanine prevented the loss of activity toward both peptide and protein
Mariangela Ceruso et al.
Biochimica et biophysica acta, 1824(2), 303-310 (2011-11-01)
The chemical shift of the carboxylate carbon of Z-tryptophan is increased from 179.85 to 182.82 ppm and 182.87 ppm on binding to thermolysin and stromelysin-1 respectively. The chemical shift of Z-phenylalanine is also increased from 179.5 ppm to 182.9 ppm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service