Skip to Content
Merck
All Photos(3)

Key Documents

359769

Sigma-Aldrich

trans,trans-2,4-Hexadienoic acid potassium salt

ReagentPlus®, 99%

Synonym(s):

Potassium sorbate, Potassium 2,4-hexadienoate, Sorbic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
Beilstein:
5357554
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

SMILES string

[K+].C\C=C\C=C\C([O-])=O

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sorbates have been reported to be less toxic than benzoate and have been classified as “Generally Recognized as Safe” (GRAS) additives by the U.S. Food and Drug Administration (FDA).Sorbic acid is metabolised to mainly to carbon dioxide. While the minor amounts are converted to trans,trans-muconic acid (ttMA), which is excreted unchanged into the urine. Urinary ttMA is a biomarker for the occupational and environmental exposure to benzene. Ability of trans,trans-2,4-Hexadienoic acid potassium salt (Sorbic acid potassium salt, Potassium sorbate) to induce chromosome aberrations, sister chromatid exchanges (SCE) and gene mutations in cultured Chinese hamster V79 cells has been examined.Potassium sorbate is reported to be less genotoxic than the sodium salt analog.

Application

  • Determination of preservatives in canned food products: Investigates the presence of preservatives, including trans,trans-2,4-Hexadienoic acid potassium salt, in canned foods (AM Charles, 2014).
  • The effect of selected preservatives on the growth of Listeria monocytogenes in ready-to-eat foods: Examines the inhibitory effects of potassium sorbate (trans-2,4-hexadienoic acid potassium salt) on Listeria monocytogenes (L Necidová, B Mrňousová, D Haruštiaková, Š Bursová, 2019).
  • Effect of leave‐on cosmetic antimicrobial preservatives on healthy skin resident Staphylococcus epidermidis: Evaluates the impact of various preservatives, including trans,trans-2,4-Hexadienoic acid potassium salt, on skin microbiota (W Zhang, X Wang, L Zhao, Y Gu, 2023).
  • Catalytic Strategy for Conversion of Triacetic Acid Lactone to Potassium Sorbate: Describes a method for synthesizing potassium sorbate from triacetic acid lactone using a catalytic process (MS Kim, D Choi, J Ha, K Choi, JH Yu, JA Dumesic, 2023).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Teresa Pérez-Prior et al.
Journal of agricultural and food chemistry, 56(24), 11824-11829 (2008-12-05)
Sorbic acid reacts with nitrite to yield mutagenic products such as 1,4-dinitro-2-methylpyrrole (NMP) and ethylnitrolic acid (ENA). In order to know the stability of these compounds, a kinetic study of their decomposition reactions was performed in the 6.0-9.5 pH range.
M M Hasegawa et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 22(7), 501-507 (1984-07-01)
The ability of sorbic acid and its potassium and sodium salts to induce chromosome aberrations, sister chromatid exchanges (SCE) and gene mutations in cultured Chinese hamster V79 cells was examined. Sodium sorbate caused significant induction of chromosome aberrations and SCE
T Renner et al.
Journal of chromatography. A, 847(1-2), 127-133 (1999-08-04)
The average daily uptake of the common food preservative sorbic acid is estimated to range from 0.01 to 1.1 mg kg-1. Sorbic acid mainly is metabolised to carbon dioxide. Minor amounts are converted to trans,trans-muconic acid (ttMA) as well as
Cheng-An Hwang et al.
Journal of food protection, 78(6), 1154-1160 (2015-06-04)
The surfaces of ready-to-eat meats are susceptible to postprocessing contamination by Listeria monocytogenes. This study quantified the lag-phase durations (LPD) and growth rates (GR) of L. monocytogenes on the surfaces of cooked ham as affected by sorbate solutions of different
Rui Cai et al.
International journal of food microbiology, 214, 145-150 (2015-08-25)
Alicyclobacillus acidoterrestris can survive the pasteurization process, multiply in pasteurized juices and produce guaiacol which causes medicinal or antiseptic off-flavors. Chemical preservatives have the potential to suppress outgrowth of surviving populations during subsequent storage of fruit juices. In the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service