341746
Copper(II) acetate hydrate
98%
Synonym(s):
Cupric acetate hydrate
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
Cu(CO2CH3)2 · xH2O
CAS Number:
Molecular Weight:
181.63 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor density
6.9 (vs air)
Assay
98%
form
crystals
composition
Degree of hydration, ~1
reaction suitability
core: copper
SMILES string
O.CC(=O)O[Cu]OC(C)=O
InChI
1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2
InChI key
NWFNSTOSIVLCJA-UHFFFAOYSA-L
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pundalik D Walke et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-08-30)
Memristive systems can provide a novel strategy to conquer the von Neumann bottleneck by evaluating information where data are located in situ. To meet the rising of artificial neural network (ANN) demand, the implementation of memristor arrays capable of performing
Anaisa A Leyva-Diaz et al.
Journal of animal science and biotechnology, 12(1), 23-23 (2021-02-06)
Interest in the use of natural feed additives as an alternative to antimicrobials in the poultry industry has increased in recent years because of the risk of bacterial resistance. One of the most studied groups are polyphenolic compounds, given their
Alexander Navarrete et al.
Faraday discussions, 183, 249-259 (2015-09-24)
A novel plasmonic reactor concept is proposed and tested to work as a visible energy harvesting device while allowing reactions to transform CO2 to be carried out. Particularly the reverse water gas shift (RWGS) reaction has been tested as a
Monica L Ohnsorg et al.
Langmuir : the ACS journal of surfaces and colloids, 31(22), 6114-6121 (2015-05-29)
Thin films can integrate the versatility and great potential found in the emerging field of metal-organic frameworks directly into device architectures. For fabrication of smart interfaces containing surface-anchored metal-organic frameworks, it is important to understand how the foundational layers form
Łukasz Orzeł et al.
Dalton transactions (Cambridge, England : 2003), 44(13), 6012-6022 (2015-02-28)
The nature of chlorophyll interactions with copper(II) ions varies considerably in organic solvents, depending on the dominant coordinative form. Besides formation of the metallo tetrapyrrolic complex, Cu(II) ions can cause oxidation of the pigment, reversible or irreversible, which can lead
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service