Skip to Content
Merck
All Photos(1)

Key Documents

341568

Sigma-Aldrich

Benzeneruthenium(II) chloride dimer

Synonym(s):

(Benzene)dichlororuthenium dimer, Benzeneruthenium dichloride dimer, Bis(benzene)tetrachlorodiruthenium, Bis[(benzene)dichlororuthenium], Dibenzenetetrachlorodiruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru2(C6H6)2Cl4
CAS Number:
Molecular Weight:
500.18
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

Cl[Ru]Cl.Cl[Ru]Cl.c1ccccc1.c2ccccc2

InChI

1S/2C6H6.4ClH.2Ru/c2*1-2-4-6-5-3-1;;;;;;/h2*1-6H;4*1H;;/q;;;;;;2*+2/p-4

InChI key

YGXMUPKIEHNBNQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Hydrosilylation Catalysts

  • Catalyst for SNAr reactions of fluoroarenes and cyclic amines (eq. 1)
  • Precatalyst for the hydrogenation of β-keto esters (eq. 2)
  • Precursor for the preparation of other ruthenium catalysts (eq. 3)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service