333468
Silicon tetrabromide
99%
Synonym(s):
Tetrabromosilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
vapor density
2.8 (vs air)
Quality Level
Assay
99%
form
liquid
reaction suitability
core: silicon
bp
153 °C (lit.)
mp
5 °C (lit.)
density
2.8 g/mL at 25 °C (lit.)
SMILES string
Br[Si](Br)(Br)Br
InChI
1S/Br4Si/c1-5(2,3)4
InChI key
AIFMYMZGQVTROK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Silicon tetrabromide (SiBr4) may be used to prepare photoluminescent silicon nanocrystals by chemical reduction with alkaline metals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Photoluminescent silicon nanocrystals stabilized by ionic liquid.
Journal of Nanoparticle Research, 13(5), 1971-1978 (2011)
Double stabilization of nanocrystalline silicon: a bonus from solvent.
Journal of Nanoparticle Research, 18(1), 17-17 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service