Skip to Content
Merck
All Photos(1)

Documents

327867

Sigma-Aldrich

4-Bromo-2,6-difluoroaniline

98%

Synonym(s):

(4-Bromo-2,6-difluorophenyl)amine, 2,6-Difluoro-4-bromoaniline, 4-Bromo-2,6-difluorobenzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(F)2NH2
CAS Number:
Molecular Weight:
208.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

63-65 °C (lit.)

SMILES string

Nc1c(F)cc(Br)cc1F

InChI

1S/C6H4BrF2N/c7-3-1-4(8)6(10)5(9)2-3/h1-2H,10H2

InChI key

BFQSQUAVMNHOEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2,6-difluoroaniline was used in synthesis of:
  • ammonium 5-bromo-7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate, fluorogenic derivatizating reagent for the determination of aminothiols by HPLC
  • olefin tolanes
  • 1-(4-amino-3,5-difluoro)-2-(4-butyloxyphenyl)ethyne
  • 4′′-n-butyloxy-3,5-difluoroterphenyl-4-amine
  • 4-(3-methylbut-1-yn-3-ol)-2,6-difluoroaniline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Giorgio Cevasco et al.
Journal of chromatography. A, 1217(14), 2158-2162 (2010-02-26)
Altered levels of aminothiols in biological fluids are thought to be an important risk indicator for several diseases, and reliable methods for the accurate determination of aminothiols concentrations in plasma are thus required. In this paper ammonium 5-bromo-7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate (SBD-BF) is
Tailoring the physical properties of some high birefringence isothiocyanato-based liquid crystals.
Catanescu CO, et al.
Liq. Cryst., 31(4), 541-555 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service