Skip to Content
Merck
All Photos(2)

Documents

325430

Sigma-Aldrich

Triphenylmethylamine

99%

Synonym(s):

Tritylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CNH2
CAS Number:
Molecular Weight:
259.34
Beilstein:
2113674
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

223 °C/14 mmHg (lit.)

mp

102-104 °C (lit.)

SMILES string

NC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H17N/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,20H2

InChI key

BZVJOYBTLHNRDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reaction of triphenylmethylamine with borontrifluoride was studied.

Application

Triphenylmethylamine was used to prepare diamondoid porous organic salt. It was also used in preparation of N-tritylated β-aminoalcohols, useful building blocks in organic synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tritylamine (triphenylmethylamine) in organic synthesis; II. The reaction of tritylamine with oxiranes-synthesis of N-trityl-? aminoalcohols.
Soroka M and Goldeman W.
ARKIVOC (Gainesville, FL, United States), 12, 31-37 (2003)
Atsushi Yamamoto et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(9), 3006-3016 (2013-01-12)
A diamondoid porous organic salt (d-POS) composed of 8-hydroxyquinoline-5-sulfonic acid (HQS) and triphenylmethylamine (TPMA) shows reversible structure contraction and expansion ("breathing") in response to guest desorption and adsorption. This flexible structure is designed hierarchically by utilizing two different types of
Reaction of triphenylmethylamines with boron trihalides.
Ronan RJ, et al.
Journal of the American Chemical Society, 93(25), 6811-6814 (1971)
Grant A McNaughton-Smith et al.
Journal of medicinal chemistry, 51(4), 976-982 (2008-02-01)
Sickle cell disease (SCD) is a hereditary condition characterized by deformation of red blood cells (RBCs). This phenomenon is due to the presence of abnormal hemoglobin that polymerizes upon deoxygenation. This effect is exacerbated when dehydrated RBCs experience a loss
Silke B Bodendiek et al.
Frontiers in pharmacology, 3, 106-106 (2012-06-12)
The paucity of specific pharmacological agents has been a major impediment for delineating the roles of gap junction (GJ) channels formed by connexin proteins in physiology and pathophysiology. Here, we used the selective optimization of side activities (SOSA) approach, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service