Skip to Content
Merck
All Photos(1)

Key Documents

324655

Sigma-Aldrich

4-tert-Butylphenylmagnesium bromide solution

2.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4MgBr
CAS Number:
Molecular Weight:
237.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

1.065 g/mL at 25 °C

SMILES string

CC(C)(C)c1ccc([Mg]Br)cc1

InChI

1S/C10H13.BrH.Mg/c1-10(2,3)9-7-5-4-6-8-9;;/h5-8H,1-3H3;1H;/q;;+1/p-1

InChI key

YECFWFOEQLAVNH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service