Skip to Content
Merck
All Photos(4)

Documents

316512

Sigma-Aldrich

Lead(II) acetate trihydrate

99.999% trace metals basis

Synonym(s):

Lead diacetate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

reaction suitability

core: lead

mp

75 °C (dec.) (lit.)

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead acetate trihydrate appears as white crystals that melt at 75 °C. It is water soluble and stable under normal conditions.

Application

Lead(II) acetate trihydrate was a component of the solution which was used towards the fabrication of platinum black microelectrodes. Lead acetate trihydrate may be used in the manufacturing of other lead salts. It may be used in the textile industry for dyeing and printing. In paint industry, lead acetate acts as a drying agent and as a purification agent in chemical synthesis.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raeyoung Kim et al.
Journal of neural engineering, 12(2), 026010-026010 (2015-03-05)
Platinum black (PtBK) has long been used for microelectrode fabrication owing to its high recording performance of neural signals. The porous structure of PtBK enlarges the surface area and lowers the impedance, which results in background noise reduction. However, the
Mi Jin Kim et al.
International journal of molecular sciences, 22(4) (2021-03-07)
Mercury is one of the detrimental toxicants that can be found in the environment and exists naturally in different forms; inorganic and organic. Human exposure to inorganic mercury, such as mercury chloride, occurs through air pollution, absorption of food or
Hazardous Chemicals: Safety Management and Global Regulations null
Electrochemical layer-by-layer approach to fabricate mechanically stable platinum black microelectrodes using a mussel-inspired polydopamine adhesive.
Kim R and Nam Y
Journal of Neural Engineering, 12(2) (2015)
Carbon-13 CP/MAS NMR and crystallographic investigations of the structure and solid-state transformations of lead (II) acetate trihydrate.
Bryant RG, et al.
Inorganic Chemistry, 23(22), 3580-3584 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service