310743
1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
functional group
sulfonic acid
SMILES string
O.[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc2c(cc(cc2c1)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C10H8O9S3.3Na.H2O/c11-20(12,13)7-1-2-9-6(3-7)4-8(21(14,15)16)5-10(9)22(17,18)19;;;;/h1-5H,(H,11,12,13)(H,14,15,16)(H,17,18,19);;;;1H2/q;3*+1;/p-3
InChI key
XJQMXRPUYORHKQ-UHFFFAOYSA-K
Application
1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate can be used as:
- An anionic chromophore in capillary electrophoresis.
- An anionic dopant for the polymerization of pyrrole.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Analytical chemistry, 85(22), 11112-11120 (2013-10-17)
New methods for quantitative analysis of strong anions are required for diagnostic testing of human diseases. Current techniques suffer from poor selectivity and/or long analysis times that are not amenable for labile anions in high-saline or volume-restricted samples. We introduce
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service