Skip to Content
Merck
All Photos(1)

Documents

306053

Sigma-Aldrich

Methylcyclohexane-d14

99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D11CD3
CAS Number:
Molecular Weight:
112.27
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

Assay

99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.4189 (lit.)

bp

101 °C

mp

-126 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

mass shift

M+14

SMILES string

[2H]C([2H])([2H])C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D

InChI key

UAEPNZWRGJTJPN-OBYKGMMLSA-N

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Origin of multiple phosphorescence emission from p-chlorobenzaldehyde in polycrystallized methylcyclohexane-d14 at 4.2 K.
Lamotte M and Goodman L.
The Journal of Physical Chemistry, 86(17), 3371-3374 (1982)
Anharmonicity and Hydrogen Bonding. The in-plane OH Bending and its Combination with the OH Stretching Vibration.
Asselin M and Sandorfy C.
Canadian Journal of Chemistry, 49(9), 1539-1544 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service