Skip to Content
Merck
All Photos(3)

Key Documents

294640

Sigma-Aldrich

(S)-4-Benzyl-2-oxazolidinone

99%

Synonym(s):

(4S)-(-)-4-(Phenylmethyl)-1,3-oxazolidin-2-one, (4S)-4-(Phenylmethyl)-2-oxazolidinone, (4S)-4-benzyl-1,3-oxazolidin-2-one, (S)-(-)-4-benzyl-2-oxazolidinone, (S)-4-benzyloxazolidin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Beilstein:
3649667
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D −63°, c = 1 in chloroform

optical purity

ee: 99% (HPLC)

mp

86-88 °C (lit.)

functional group

phenyl

SMILES string

O=C1N[C@H](CO1)Cc2ccccc2

InChI

1S/C10H11NO2/c12-10-11-9(7-13-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,11,12)/t9-/m0/s1

InChI key

OJOFMLDBXPDXLQ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral auxiliary for asymmetric alkylation.
Chiral auxiliary used recently in a synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides.
Used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert K Boeckman et al.
The Journal of organic chemistry, 71(23), 8969-8972 (2006-11-04)
Camphor-derived lactams and other related chiral controllers have been found to react with the Bestmann ylide (triphenylphosphoranylideneketene) upon heating in toluene. The resulting parent ylides provide convenient access to a structurally diverse set of chiral stabilized ylides via functionalization. The
Jennifer M Heemstra et al.
Organic letters, 8(24), 5441-5443 (2006-11-17)
The first enantioselective total synthesis of (S)-equol is reported. The described route relies on an Evans alkylation to form the stereocenter and an intramolecular Buchwald etherification to generate the chroman ring. Key features of this method include its brevity, its
Arun K Ghosh et al.
Journal of the American Chemical Society, 126(12), 3704-3705 (2004-03-25)
An enantioselective first total syntheis of amphidinolide W (2) and a revision of its C6 absolute stereochemistry (1) are described. Amphidinolide W (1), a 12-membered macrolide isolated from Amphidinium sp., has shown potent antitumor properties against a variety of NCI
Tetrahedron Letters, 33, 7613-7613 (1992)
Anne N Shemon et al.
PloS one, 4(6), e6028-e6028 (2009-06-25)
Raf Kinase Inhibitory Protein (RKIP, also PEBP1), a member of the Phosphatidylethanolamine Binding Protein family, negatively regulates growth factor signaling by the Raf/MAP kinase pathway. Since an organic compound, locostatin, was reported to bind RKIP and inhibit cell migration by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service