Skip to Content
Merck
All Photos(1)

Key Documents

28684

Sigma-Aldrich

trans-Cyclobutane-1,2-dicarboxylic acid

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Beilstein:
3198993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

form

solid

SMILES string

OC(=O)[C@@H]1CC[C@H]1C(O)=O

InChI

1S/C6H8O4/c7-5(8)3-1-2-4(3)6(9)10/h3-4H,1-2H2,(H,7,8)(H,9,10)/t3-,4-/m1/s1

InChI key

SUSAGCZZQKACKE-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The interaction of the carboxyl groups in the fragmentation of the molecular ions of cis-cyclobutane-1,2-dicarboxylic acid helps to distinguish it from trans-cyclobutane-1,2-dicarboxylic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit F and Holmes JL.
Org. Mass Spectrom., 6(5), 541-548 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service