Skip to Content
Merck
All Photos(1)

Key Documents

28623

Sigma-Aldrich

4-(Heptadecafluorooctyl)aniline

≥95.0% (GC)

Synonym(s):

4-(Perfluorooctyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6F17N
CAS Number:
Molecular Weight:
511.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

solid

mp

42-47 °C

functional group

fluoro

SMILES string

NC1=CC=C(C(F)(C(F)(C(F)(C(F)(C(F)(C(F)(C(F)(C(F)(F)F)F)F)F)F)F)F)F)C=C1

InChI

1S/C14H6F17N/c15-7(16,5-1-3-6(32)4-2-5)8(17,18)9(19,20)10(21,22)11(23,24)12(25,26)13(27,28)14(29,30)31/h1-4H,32H2

InChI key

ZQTZVMUNYIDMJV-UHFFFAOYSA-N

Application

4-(Heptadecafluorooctyl)aniline was used in the synthesis of fluorous (S)-pyrrolidine-thiourea bifunctional organocatalyst.

Other Notes

Building block for introducing a 4-perfluoroalkylaryl moiety

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Matsui et al.
Liq. Cryst., 21, 669-669 (1996)
N. Yoshino et al.
Bulletin of the Chemical Society of Japan, 65, 2141-2141 (1992)
D. Crich et al.
Tetrahedron, 55, 14261-14261 (1999)
Liang Wang et al.
Synlett : accounts and rapid communications in synthetic organic chemistry, 3(2010), 433-436 (2010-08-19)
A novel fluorous (S)-pyrrolidine-thiourea bifunctional organocatalyst is prepared. The catalyst shows good activity and enantioselectivity for direct α-chlorination of aldehydes using N-chlorosuccinimide (NCS) as the chlorine source. It can be recovered from the reaction mixture by fluorous solid-phase extraction with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service