Skip to Content
Merck
All Photos(1)

Key Documents

283509

Sigma-Aldrich

4-Ethylbenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4COCl
CAS Number:
Molecular Weight:
168.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5479 (lit.)

bp

235-236 °C (lit.)

density

1.147 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCc1ccc(cc1)C(Cl)=O

InChI

1S/C9H9ClO/c1-2-7-3-5-8(6-4-7)9(10)11/h3-6H,2H2,1H3

InChI key

AVTLLLZVYYPGFX-UHFFFAOYSA-N

Related Categories

Application

4-Ethylbenzoyl chloride has been used in the preparation of:
  • N-benzoyl-4-ethylbenzamide
  • N-(4-diethylaminobenzoyl)-2-methoxybenzenesulfonamide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina Fomovska et al.
Antimicrobial agents and chemotherapy, 56(5), 2666-2682 (2012-02-23)
Toxoplasma gondii is a protozoan parasite that can damage the human brain and eyes. There are no curative medicines. Herein, we describe our discovery of N-benzoyl-2-hydroxybenzamides as a class of compounds effective in the low nanomolar range against T. gondii

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service