Skip to Content
Merck
All Photos(4)

Key Documents

267953

Sigma-Aldrich

Iron

powder, <10 μm, ≥99.9% trace metals basis

Synonym(s):

Fe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
11101713
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

core: iron
reagent type: catalyst

resistivity

9.71 μΩ-cm

particle size

<10 μm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Application

Iron powder is commonly used as the catalyst for a wide variety of organic transformations. Applications include:
  • Reduction of nitroaromatic compounds to aromatic amines.[1][2]
  • Iron-mediated polymerization of methyl methacrylate (MMA) in the presence of a reversible addition-fragmentation chain transfer (RAFT) agent, 2-cyanoprop-2-yl 1-dithionaphthalate (CPDN).[3]
  • Synthesis of pyrrolo[1,2-a]quinoxaline derivatives.[4]
  • Conversion of ketoximes to the corresponding ketones by treatment with iron powder.[5]
  • Iron-catalyzed cross-coupling reaction of imidazoles with vinyl bromides and vinyl chlorides.[6]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 7

1 of 7

Krafft, M. E.; Holton, R. A.
Journal of the American Chemical Society, 106, 7619-7619 (1984)
Ito, S.; Inoue, K.; Mastumoto, M.
Journal of the American Chemical Society, 104, 6450-6450 (1982)
One-pot synthesis of pyrrolo [1, 2-a] quinoxaline derivatives via iron-promoted aryl nitro reduction and aerobic oxidation of alcohols.
Pereira M D F and Thiery V
Organic Letters, 14(18), 4754-4757 (2012)
Reduction of nitroarenes to aromatic amines with nanosized activated metallic iron powder in water.
Wang L, et al.
Synthesis, 2003(13), 2001-2004 (2003)
Iron?Catalyzed Cross?Coupling Reaction of Vinyl Bromides or Chlorides with Imidazoles in the Absence of Ligands and Additives.
Mao J, et al.
Advanced Synthesis & Catalysis, 351(9), 1268-1272 (2009)

Questions

  1. Do you supply assay with your iron, what is size range of powder, morphology etc

    1 answer
    1. The particle morphology is not determined. The minimum purity of this material is 99.9% and the maximum particle size is 10 um. This information is lot specific and reported in the Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/aldrich/267953#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service