266957
Aluminum
foil, thickness 1.0 mm, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.999% trace metals basis
form
foil
autoignition temp.
1400 °F
resistivity
2.6548 μΩ-cm
thickness
1.0 mm
bp
2460 °C (lit.)
mp
660.37 °C (lit.)
density
2.7 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Related Categories
Quantity
100 x 100 mm (approximately 27.2 g)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate
Caitlin Carney et al.
Journal of the American Academy of Dermatology, 69(4), 578-582 (2013-07-17)
Laser treatment has emerged as a novel treatment modality for onychomycosis. We sought to determine thermal response and optical effects of a submillisecond neodymium:yttrium-aluminum-garnet (Nd:YAG) 1064-nm laser on common fungal nail pathogens, and the clinical efficacy and safety of the
Jesper C Holst et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8819-8823 (2013-05-15)
Refractory inclusions [calcium-aluminum-rich inclusions, (CAIs)] represent the oldest Solar System solids and provide information regarding the formation of the Sun and its protoplanetary disk. CAIs contain evidence of now extinct short-lived radioisotopes (e.g., (26)Al, (41)Ca, and (182)Hf) synthesized in one
Arjun Joshua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), 9633-9638 (2013-05-28)
Controlling the coupling between localized spins and itinerant electrons can lead to exotic magnetic states. A novel system featuring local magnetic moments and extended 2D electrons is the interface between LaAlO3 and SrTiO3. The magnetism of the interface, however, was
Mutlu Ozcan et al.
The journal of adhesive dentistry, 15(3), 211-214 (2013-05-24)
To evaluate the effect of nozzle distance, nozzle angle, and deposition duration on the silica content attained on zirconia by air abrasion. Disk-shaped zirconia (LAVA, 3M ESPE) (diameter: 10 mm, thickness: 2 mm) specimens (N = 54) were obtained. They
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service