Skip to Content
Merck
All Photos(2)

Documents

266167

Sigma-Aldrich

Manganese

chips, thickness <2.0 mm, 99%

Synonym(s):

Manganese element, Manganese-55

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

chips

reaction suitability

reagent type: catalyst
core: manganese

resistivity

185 μΩ-cm, 20°C

thickness

<2.0 mm

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Barceloux
Journal of toxicology. Clinical toxicology, 37(2), 293-307 (1999-06-26)
Manganese is a very hard, brittle metal, which is used to increase the strength of steel alloys. Absorption from the gastrointestinal tract occurs in the divalent and tetravalent forms. Permanganates, which are strong oxidizing agents, have a +7 valence. The
Alan Woolf et al.
Environmental health perspectives, 110(6), 613-616 (2002-06-11)
The patient's family bought a home in a suburb, but the proximity of the house to wetlands and its distance from the town water main prohibited connecting the house to town water. The family had a well drilled and they
M Baldwin et al.
Neurotoxicology, 20(2-3), 343-353 (1999-07-01)
Exposure data and bioindicators were obtained for a study whose objective was detection of early manifestations of manganese (Mn) neurotoxicity in a population with potential environmental exposure. The study included persons with no history of neurotoxic workplace exposure in Southwest
D Mergler et al.
Neurotoxicology, 20(2-3), 327-342 (1999-07-01)
Excessive manganese (Mn) has been associated with neurobehavioral deficits and neurological and/or neuropsychiatric illness, but the level at which this metal can cause adverse neurotoxic effects, particularly with long-term exposure, is still unknown. The objective of the present study was
Rachel J Service et al.
Biochemistry, 52(47), 8452-8464 (2013-10-31)
In the 1.9 Å structural model of photosystem II (PDB: 3ARC), the amino acid residue Glu333 of the D1 polypeptide coordinates to the oxygen-evolving Mn₄CaO₅ cluster. This residue appears to be highly significant in that it bridges the two Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service