Skip to Content
Merck
All Photos(1)

Key Documents

260282

Sigma-Aldrich

1,2-Pentanediol

96%

Synonym(s):

1, 2-Dihydroxypentane, 1, 2-Pentylene glycol, 4-Methyl-1 2-butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1732199
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

206 °C (lit.)

density

0.971 g/mL at 25 °C (lit.)

SMILES string

CCCC(O)CO

InChI

1S/C5H12O2/c1-2-3-5(7)4-6/h5-7H,2-4H2,1H3

InChI key

WCVRQHFDJLLWFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Pentanediol belongs to the class of diol monomers commonly used in the preparation of various polymers including polyesters and polyurethanes. These polymers are used in packaging, engineering plastic, textiles, cosmetics, detergents, lubricants, optical films, polyester resins, polyurethane foams, inks, adhesives, and sealants due to their desirable properties such as flexibility, durability, chemical resistance, and thermal insulation properties. 1,2-Pentanediol also finds diverse applications, serving as an organic solvent, plasticizer, surfactant, and reactant in the synthesis of various compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyo Shigeyama et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1104, 49-58 (2018-11-18)
In recent years, volatile organic compounds (VOCs) discharged from the human body, of which some compounds exhibit strong correlations with pathological conditions, have attracted attention as a new means of disease diagnosis technology. The aim of this study was to
Miguel Paz-Alvarez et al.
International journal of pharmaceutics, 549(1-2), 317-324 (2018-07-29)
Dandruff is a common condition, affecting up to half the global population of immunocompetent adults at some time during their lives and it has been highly correlated with the over-expression of the fungus Malassezia spp. Climbazole (CBZ) is used as
Richard Krombholz et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
In the development of dermal drug formulations and cosmetics, understanding the penetration properties of the active ingredients is crucial. Given that widespread methods, including tape stripping, lack in spatial resolution, while being time- and labour-intensive, Confocal Raman Microscopy is a
Kan Shoji et al.
ACS nano, 13(2), 2606-2614 (2019-02-07)
In this article, we describe the quantitative characterization of a gold nanoneedle ion channel probe and demonstrate the utility of this probe for spatially resolved detection of a small molecule using ion channel activity. Our report builds on recent reports
Anna-Carola Eicher et al.
International journal of pharmaceutics, 563, 208-216 (2019-04-07)
Biopharmaceutical development is progressing rapidly. It is imperative that novel drug delivery systems are designed to protect the integrity of the biopharmaceutical, and, at the same time, transport and distribute the drug efficaciously to the target site. Administration of highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service