Skip to Content
Merck
All Photos(1)

Key Documents

259519

Sigma-Aldrich

3-Nonanone

99%

Synonym(s):

Ethyl hexyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COC2H5
CAS Number:
Molecular Weight:
142.24
Beilstein:
1700447
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.42 (lit.)

bp

187-188 °C (lit.)

density

0.821 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCCCCCC(=O)CC

InChI

1S/C9H18O/c1-3-5-6-7-8-9(10)4-2/h3-8H2,1-2H3

InChI key

IYTXKIXETAELAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-Nonanone has been used as internal standard in determination of significant volatile components of pacharan (alcoholic beverage).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of significant volatile components of pacharan.
Fernandez-Garcia T, et al.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 206(6), 414-416 (1998)
Peter S Toth et al.
The Analyst, 140(6), 1947-1954 (2015-02-11)
Two-phase voltammetry has been carried out using a reverse cell configuration, i.e. with the lower density organic solvent on the top of the aqueous solution in the cell, where the organic solvents contain either nitrile or ketone functional groups. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service