Skip to Content
Merck
All Photos(3)

Key Documents

257303

Sigma-Aldrich

1,2,3,4-Butanetetracarboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(CO2H)CH2CO2H]2
CAS Number:
Molecular Weight:
234.16
Beilstein:
1729167
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

195-197 °C (dec.) (lit.)

SMILES string

OC(=O)CC(C(CC(O)=O)C(O)=O)C(O)=O

InChI

1S/C8H10O8/c9-5(10)1-3(7(13)14)4(8(15)16)2-6(11)12/h3-4H,1-2H2,(H,9,10)(H,11,12)(H,13,14)(H,15,16)

InChI key

GGAUUQHSCNMCAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,3,4-Butanetetracarboxylic acid (BTCA) is utilized as a cross-linking agent and as a building block for the synthesis of specialized polymers for the formulation of resins, polymers, coatings, and adhesives.

Application

1,2,3,4-Butanetetracarboxylic acid (BTCA) can be used as a cross-linking agent:
  • To functionalize cotton fabric. BTCA-treated fabric shows improved anti-pilling, wrinkle resistance, and fire-retardant properties.
  • To fabricate flexible, free-standing nanocellulose membranes. The cross-linking with BTCA improves water stability and ionic conductivity of membranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asli Celebioglu et al.
Scientific reports, 7(1), 7369-7369 (2017-08-09)
Water pollution is a serious concern for public health and environment in today's world; hence, there exists a strong demand to develop cost-effective, sustainable and eco-friendly membranes. Here, we produce a highly efficient molecular filter membrane based on bio-renewable material;
Bolin Ji et al.
Carbohydrate polymers, 207, 288-296 (2019-01-03)
The sodium l-glutamate is reported as an efficient catalyst for the cross-linking between 1,2,3,4-butanetetracarboxylic acid (BTCA) and cellulose. Results presented ester absorbance of the treated fabrics strongly increased in the presence of the homemade l-glutamate salt, a mixture of l-glutamic
Peipei Wang et al.
Carbohydrate polymers, 218, 103-111 (2019-06-22)
Environmentally friendly, sustainable, and high-performance thermal insulators are in high demand. Petroleum-based insulator foams usually have high thermal conductivity and pose health hazards. Here, we report ultralight composite foams that are highly strong, elastic, and super-insulating. The foams are composed
Solmaz Heydarifard et al.
Carbohydrate polymers, 181, 1086-1092 (2017-12-20)
Development of a foam-formed cellulose filter paper with high wet strength was carried out for application as a drinking water filter. The wet strength and antimicrobial activity of cellulose foam paper against several bacteria species (Bacillus subtilis MTCC 441 (Gram
Eduardo de O Sanguinet et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(3), 857-867 (2019-06-30)
Induced pluripotent stem cells (iPSCs) provide a promising means of creating custom-tailored cell lines for cellular therapies. Their application in regenerative medicine, however, depends on the possibility that the maintenance and differentiation of cells and organs occur under defined conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service