Skip to Content
Merck
All Photos(3)

Key Documents

255696

Sigma-Aldrich

Calcium sulfate

≥99.99% trace metals basis

Synonym(s):

Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

≥99.99% trace metals basis

form

powder and chunks

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium sulfate (CaSO4) is an inorganic compound with significant applications in material science, particularly due to its various hydrated forms, such as gypsum and plaster of Paris. It can be used in the production of solar thermal energy systems, where it serves as a heat transfer medium or in the construction of solar panels. Additionally, calcium sulfate can also be utilized in the formulation of electrolytes for calcium-ion batteries.

Application

Calcium sulfate can be used:   
  • In the formulation of electrolytes for calcium-ion batteries. The presence of calcium sulfate helps in improving the solubility of calcium salts in the electrolyte.      
  • As a surface modifier to enhance the performance of Ni-rich lithium nickel-cobalt-manganese oxide (NCM) cathode materials in lithium-ion batteries.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Gholami et al.
Chemosphere, 212, 105-113 (2018-08-26)
This study investigated the applicability of synthesized calcium peroxide (CaO2) nanoparticles for naphthalene bioremediation by permeable reactive barrier (PRB) from groundwater. According to the batch experiments the application of 400 mg/L of CaO2 nanoparticles was the optimum concentration for naphthalene (20 mg/L)
Vincoli JW
Risk Management for Hazardous Chemicals null
Claudia Fischbach et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(2), 399-404 (2009-01-08)
Three-dimensional culture alters cancer cell signaling; however, the underlying mechanisms and importance of these changes on tumor vascularization remain unclear. A hydrogel system was used to examine the role of the transition from 2D to 3D culture, with and without
Eoin McEvoy et al.
Nature communications, 11(1), 6148-6148 (2020-12-03)
Sustained proliferation is a significant driver of cancer progression. Cell-cycle advancement is coupled with cell size, but it remains unclear how multiple cells interact to control their volume in 3D clusters. In this study, we propose a mechano-osmotic model to
Kathryn A Dooley et al.
The Analyst, 138(17), 4838-4848 (2013-06-27)
In situ chemical imaging techniques are being developed to provide information on the spatial distribution of artists' pigments used in polychrome works of art such as paintings. The new methods include reflectance imaging spectroscopy and X-ray fluorescence mapping. Results from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service