Skip to Content
Merck
All Photos(1)

Key Documents

253596

Sigma-Aldrich

16-DOXYL-stearic acid, free radical

Synonym(s):

16-SASL, 16DOXYL, 16NS, 2-(14-Carboxytetradecyl)-2-ethyl-4,4-dimethyl-3-oxazolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42NO4
CAS Number:
Molecular Weight:
384.57
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

47-55 °C (lit.)

functional group

carboxylic acid
ether

storage temp.

2-8°C

SMILES string

CCC1(CCCCCCCCCCCCCCC(O)=O)OCC(C)(C)N1[O]

InChI

1S/C22H42NO4/c1-4-22(23(26)21(2,3)19-27-22)18-16-14-12-10-8-6-5-7-9-11-13-15-17-20(24)25/h4-19H2,1-3H3,(H,24,25)

InChI key

RPAZYIOIDZRJOO-UHFFFAOYSA-N

Application

Involved in membrane studies including:
  • Dynamics and state of lipid bilayer-internal water using 1H dynamic nuclear polarization
  • Distribution of fatty acids
  • Transversal relaxation of nitroxides for PELDOR Spectroscopy
  • Effects of different lipid components on detergent-resistant membranes
Spin label for membrane fluidity studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Qi et al.
Circulation, 104(13), 1544-1549 (2001-09-26)
The maintenance and termination of reentry arrhythmias are determined by tissue properties such as refractoriness and conduction velocity. Although the effects of Na(+) and K(+) channel block on electrophysiological properties and defibrillation threshold (DFT) have been studied, little is known
Gábor Balogh et al.
PloS one, 6(6), e21182-e21182 (2011-06-24)
Cellular membranes respond rapidly to various environmental perturbations. Previously we showed that modulations in membrane fluidity achieved by heat stress (HS) resulted in pronounced membrane organization alterations which could be intimately linked to the expression and cellular distribution of heat
M A Baqué et al.
Biochemical and biophysical research communications, 505(4), 1038-1042 (2018-10-13)
Different spin labels were incorporated to the membranes of cultured insect UFL-AG-286 cells in order to characterize their physical properties by Electron Paramagnetic Resonance spectroscopy (EPR). The spectrum of the spin label 12-SASL incorporated to cell membranes was similar as those
P V Dubovskii et al.
Journal of molecular biology, 305(1), 137-149 (2000-12-15)
Carditoxins (CTXs) from cobra snake venoms, the basic 60-62 residue all-beta sheet polypeptides, are known to bind to and impair the function of cell membranes. To assess the membrane induced conformation and orientation of CTXs, the interaction of the P-type
K Tsuda et al.
Biochemical and biophysical research communications, 275(3), 946-954 (2000-09-07)
It has been shown that rheological abnormality might be an etiological factor in hypertension. Recent studies have revealed that human erythrocytes possess a nitric oxide (NO) synthase and that this activation might be involved in the regulation of rheological properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service