Skip to Content
Merck
All Photos(1)

Documents

252123

Sigma-Aldrich

4-Iodobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4COCl
CAS Number:
Molecular Weight:
266.46
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

120-121 °C/1 mmHg (lit.)

mp

63-65 °C (lit.)

SMILES string

ClC(=O)c1ccc(I)cc1

InChI

1S/C7H4ClIO/c8-7(10)5-1-3-6(9)4-2-5/h1-4H

InChI key

NJAKCIUOTIPYED-UHFFFAOYSA-N

Application

4-Iodobenzoyl chloride has been used in:
  • modification of poly(allylamine), to make the polymer x-ray visible
  • preparation of series of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists
  • preparation of [2] rotaxane monomer, for poly [2] rotaxane synthesis
  • synthesis of pyrroles from imines and acetylenes mediated by isocyanides versus palladium catalysis

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian Egle et al.
Bioorganic & medicinal chemistry letters, 14(19), 4847-4850 (2004-09-03)
A series of N-(1-benzylpyrrolidin-3-yl)arylbenzamides 8 has been prepared, and their structure-activity relationships studied. Potent ligands selective for human D(4) (hD(4)) over hD(2) and alpha(1) have been identified. One example was determined to be an antagonist in a cAMP assay, with
Synthesis of poly [2] rotaxane by Sonogashira polycondensation.
Sasabe H, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 45(17), 4154-4160 (2007)
Damia Mawad et al.
Nanotechnology, 21(33), 335603-335603 (2010-07-27)
Contrast agents are currently used in a variety of diagnostic imaging techniques, including computer tomography for early cancer detection. Radiopaque nanoparticles have recently been proposed as an alternative method to traditional contrast agents that may allow for long-term image tracking.
Ni Chen et al.
Journal of Asian natural products research, 22(5), 444-451 (2019-03-20)
A series of aromatic or long-chain chrysin derivatives (1-10) were synthesized by esterification of chrysin and acyl chloride. The chemical structures of these compounds were determined by mass spectrum (MS), 1H NMR, and 13C NMR spectra. Though aromatic chrysin derivatives
Daniel J St Cyr et al.
Organic letters, 9(3), 449-452 (2007-01-26)
[reaction: see text] A direct synthesis of pyrroles from imines, acid chlorides, and alkynes mediated by isocyanides is reported. This reaction proceeds with a range of each of these three substrates, providing a method to generate families of pyrroles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service