Skip to Content
Merck
All Photos(3)

Documents

251054

Sigma-Aldrich

2-Hydroxypyridine-3-carboxylic acid

98%

Synonym(s):

2-Hydroxynicotinic acid, 2-Hydroxypyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
Beilstein:
472167
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

258-261 °C (lit.)

solubility

formic acid: soluble 50 mg/mL, clear, colorless to yellow

SMILES string

OC(=O)c1cccnc1O

InChI

1S/C6H5NO3/c8-5-4(6(9)10)2-1-3-7-5/h1-3H,(H,7,8)(H,9,10)

InChI key

UEYQJQVBUVAELZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Hydroxypyridine-3-carboxylic acid may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anita V Thomae et al.
Pharmaceutical research, 24(8), 1457-1472 (2007-03-28)
Lipid bilayers regulate the passage of solutes into and between cellular compartments. A general prerequisite for this passage is the partitioning of the solute into the bilayer. We investigated the relationship between bilayer partitioning and permeation of three drug-like acids
Anita V Thomae et al.
Biophysical journal, 89(3), 1802-1811 (2005-06-14)
According to the pH-partition hypothesis the charged species of organic compounds do not contribute to lipid bilayer permeation as they generally show negligible partitioning into n-octanol. With this assumption, membrane permeation is related to the molar fraction of the neutral
Ade Arsianti et al.
Bioorganic & medicinal chemistry letters, 20(14), 4018-4020 (2010-07-14)
Novel 2-hydroxynicotinoyl-serine-butyl esters have been synthesized. Three-step reactions from L-serine by esterification with n-butanol, amidation with 2-hydroxynicotinic acid and esterification with the corresponding carboxylic acids gave AD-1, AD-2 and AD-3. The toxicity level of esters were determined by Brine shrimp
V B Di Marco et al.
Annali di chimica, 91(9-10), 595-603 (2002-01-05)
The formation of complexes between iron(III) and two ligands of possible relevance to chelation therapy, 2-hydroxynicotinic acid and 3-hydroxypicolinic acid, in aqueous 0.6 m (Na)Cl at 25 degrees C, has been investigated by means of potentiometric titrations and UV measurements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service