Skip to Content
Merck
All Photos(1)

Key Documents

247847

Sigma-Aldrich

2,6-Diaminopurine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N6
CAS Number:
Molecular Weight:
150.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

>300 °C (lit.)

SMILES string

Nc1nc(N)c2nc[nH]c2n1

InChI

1S/C5H6N6/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H5,6,7,8,9,10,11)

InChI key

MSSXOMSJDRHRMC-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asif Hayat et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-11)
This work incorporates a variety of conjugated donor-acceptor (DA) co-monomers such as 2,6-diaminopurine (DP) into the structure of a polymeric carbon nitride (PCN) backbone using a unique nanostructure co-polymerization strategy and examines its photocatalytic activity performance in the field of
Alexander Fuhrmann et al.
Biophysical journal, 102(10), 2381-2390 (2012-06-09)
Electron-tunneling data suggest that a noncovalently-bonded complex of three molecules, two recognition molecules that present hydrogen-bond donor and acceptor sites via a carboxamide group, and a DNA base, remains bound for seconds. This is surprising, given that imino-proton exchange rates
Ondřej Baszczyňski et al.
Bioorganic & medicinal chemistry, 19(7), 2114-2124 (2011-03-25)
An efficient method for the synthesis of N(9)-[3-fluoro-2-(phosphonomethoxy)propyl] (FPMP) derivatives of purine bases has been developed. Both (R)- and (S)-enantiomers of the N(6)-substituted FPMP derivatives of adenine and 2,6-diaminopurine were prepared and their anti-human immunodeficiency virus (HIV) and anti-Moloney murine
Takumi Ishizuka et al.
Nucleic acids research, 36(5), 1464-1471 (2008-01-22)
Invasion of two PNA strands to double-stranded DNA is one of the most promising methods to recognize a predetermined site in double-stranded DNA (PNA = peptide nucleic acid). In order to facilitate this 'double-duplex invasion', a new type of PNA
Ana María Atria et al.
Acta crystallographica. Section C, Crystal structure communications, 67(Pt 5), m169-m172 (2011-05-05)
The two isomorphous title compounds, [M(C(5)H(7)N(6))(2)(C(9)H(6)O(4))(2)(H(2)O)(2)]·4H(2)O or M(2+)(Hdap(+))(2)(hpt(2-))(2)(H(2)O)(2)·4H(2)O {where dap is 2,6-diaminopurine, H(2)hpt is homophthalic acid [2-(2-carboxyphenyl)acetic acid] and M is Ni(II) or Co(II)}, consist of neutral M(2+)(Hdap(+))(2)(hpt(2-))(2)(H(2)O)(2) monomers, where the M(II) cation lies on an inversion centre and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service