Skip to Content
Merck
All Photos(3)

Key Documents

245763

Sigma-Aldrich

Manganese(II) acetylacetonate

Synonym(s):

2,4-Pentanedione manganese(II) derivative, Manganous acetylacetonate, Mn(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O)CH3]2Mn
CAS Number:
Molecular Weight:
253.15
Beilstein:
4157965
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Mn, 21.0-23.0% EDTA titration

reaction suitability

core: manganese

mp

248-250 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Mn]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Mn/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

ZQZQURFYFJBOCE-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(II) acetylacetonate (Mn(acac)2) is a transition metal acetylacetonate that is trimeric in hydrocarbon solvents. It is monomeric and tetrahedral in its vapor state. It shows Mn-O bond energy of 65 kcal mol-1. It can be used with potassium permanganate in preparation of manganese triad (Mn(acac)3).

Application

  • Effect of Precursors on the Electrochemical Properties of Mixed RuOx/MnOx Electrodes Prepared by Thermal Decomposition.: This study investigates the impact of different precursors, including manganese(II) acetylacetonate, on the electrochemical properties of mixed RuOx/MnOx electrodes. The findings highlight the importance of precursor selection in optimizing electrode performance for energy storage applications (Petrucci et al., 2022).
  • Trinuclear coordination assemblies of low-spin dicyano manganese(II) (S = 1/2) and iron(II) (S = 0) phthalocyanines with manganese(II) acetylacetonate, tris(cyclopentadienyl)gadolinium(III) and neodymium(III).: This research explores the formation and properties of trinuclear coordination complexes involving manganese(II) acetylacetonate. These complexes exhibit unique magnetic properties that have potential applications in molecular magnetism and materials science (Romanenko et al., 2022).
  • ZIF-8 Membrane Permselectivity Modification by Manganese(II) Acetylacetonate Vapor Treatment.: This paper demonstrates how manganese(II) acetylacetonate vapor treatment can enhance the permselectivity of ZIF-8 membranes. Such modifications improve membrane performance in gas separation processes, offering advancements in industrial applications (Hayashi et al., 2021).
  • Tuning the size and composition of manganese oxide nanoparticles through varying temperature ramp and aging time.: The study focuses on the synthesis of manganese oxide nanoparticles using manganese(II) acetylacetonate as a precursor. It outlines methods to control nanoparticle size and composition, which are critical for applications in catalysis and biomedicine (Martinez de la Torre et al., 2020).
  • Manganese Oxide Nanoparticle Synthesis by Thermal Decomposition of Manganese(II) Acetylacetonate.: This research details the synthesis of manganese oxide nanoparticles via the thermal decomposition of manganese(II) acetylacetonate. The resulting nanoparticles show promise for use in various catalytic and biomedical applications (Martinez de la Torre and Bennewitz, 2020).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chemistry of Manganese, Technetium and Rhenium: Pergamon Texts in Inorganic Chemistry, 868-872 (2016)
Markus Kubin et al.
Inorganic chemistry, 57(9), 5449-5462 (2018-04-11)
The 3d transition metals play a pivotal role in many charge transfer processes in catalysis and biology. X-ray absorption spectroscopy at the L-edge of metal sites probes metal 2p-3d excitations, providing key access to their valence electronic structure, which is
Acetylacetonato manganese (III)
Charles RG and Bryant BE
Inorganic Syntheses, 7, 183-184 (1963)
Byunghoon Kang et al.
Nanomaterials (Basel, Switzerland), 9(3) (2019-03-14)
We designed a high-sensitivity magnetic resonance imaging contrast agent that could be used to diagnose diseases. First, magnetic nanocrystals were synthesized by a thermal decomposition method on an organic solvent to obtain a high magnetism and methoxy poly(ethylene glycol)-poly(lactic acid)
Zhan'ao Tan et al.
Physical chemistry chemical physics : PCCP, 14(42), 14589-14595 (2012-09-28)
A solution-processed vanadium oxide (s-VO(x)) anode buffer layer on an indium-tin-oxide (ITO) electrode was used instead of PEDOT:PSS for improving the stability and photovoltaic performance of the polymer solar cells (PSCs). The s-VO(x) layer was prepared by spin-coating a vanadyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service