Skip to Content
Merck
All Photos(2)

Documents

244503

Sigma-Aldrich

4-Methylsalicylic acid

99%

Synonym(s):

2-Hydroxy-4-methylbenzoic acid, m-Cresotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)CO2H
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

173-177 °C (lit.)

SMILES string

Cc1ccc(C(O)=O)c(O)c1

InChI

1S/C8H8O3/c1-5-2-3-6(8(10)11)7(9)4-5/h2-4,9H,1H3,(H,10,11)

InChI key

NJESAXZANHETJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The heteroleptic tris-cyclometalated iridium(III) complex with 4-methylsalicylic acid (Msal) was synthesized and characterized.

Application

4-Methylsalicylic acid was used to inhibit the medium chain acyl-CoA synthetase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-xia Guo et al.
Guang pu xue yu guang pu fen xi = Guang pu, 30(1), 170-173 (2010-03-23)
New heteroleptic tris-cyclometalated iridium(III) complexes (ppy)2 Ir(LX) (ppy==2-phenylpyridine, LX==Sal (salicylic acid), Msal (4-methylsalicylic acid), FSal(4-trifluoro methyl salicylic acid)) was synthesized and characterized. The molecular structure, photophysical properties and thermal stability were tested and analyzed. The results show that the absorption
F Kasuya et al.
Biochemical pharmacology, 52(10), 1643-1646 (1996-11-22)
Molecular characteristics of carboxylic acids were investigated for the ability to inhibit a purified medium chain acyl-CoA synthetase, using hexanoic acid as a substrate. Salicylic acid, 4-methylsalicylic acid, 2-hydroxynaphtoic acid, and 2-hydroxyoctanoic acid, which do not act as substrates for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service