Skip to Content
Merck
All Photos(1)

Key Documents

244406

Sigma-Aldrich

1-Dodecyne

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9C≡CH
CAS Number:
Molecular Weight:
166.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

215 °C (lit.)

mp

−19 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCC#C

InChI

1S/C12H22/c1-3-5-7-9-11-12-10-8-6-4-2/h1H,4-12H2,2H3

InChI key

ZVDBUOGYYYNMQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiongwu Kang et al.
Journal of the American Chemical Society, 134(3), 1412-1415 (2012-01-11)
Stable ruthenium nanoparticles were prepared by the self-assembly of 1-dodecyne onto the "bare" Ru colloid surface. The formation of a Ru-vinylidene (Ru═C═CH-R) interfacial bonding linkage was confirmed by the specific reactivity of the nanoparticles with imine derivatives to form a
Milan Milosevic et al.
Applied spectroscopy, 74(3), 305-313 (2019-11-21)
This article presents a method for extracting the optical constants of homogeneous isotropic materials using the infrared spectra of that material. The method is based on using the harmonic oscillator model of molecular polarizability to obtain optical constants, then calculating
Toshiaki Taira et al.
Journal of oleo science, 69(8), 871-882 (2020-07-10)
In this study, an N-heterocyclic carbene (NHC)-based metal coordinate surfactant (MCS), NHC-Au-MCS, in which the NHC framework afforded the bonding of the Au(I) at the linkage of the hydrophilic and hydrophobic moieties, was synthesized. The structure of NHC-Au-MCS was confirmed
Davide Albani et al.
Nature communications, 9(1), 2634-2634 (2018-07-08)
Ensemble control has been intensively pursued for decades to identify sustainable alternatives to the Lindlar catalyst (PdPb/CaCO3) applied for the partial hydrogenation of alkynes in industrial organic synthesis. Although the geometric and electronic requirements are known, a literature survey illustrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service