Skip to Content
Merck
All Photos(3)

Key Documents

241059

Sigma-Aldrich

Tetraethylammonium bromide

ReagentPlus®, 99%

Synonym(s):

TEAB, TEA bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
Beilstein:
3563430
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

crystals

impurities

1% triethylamine hydrobromide

pH

6.5 (100 g/L)

mp

285 °C (dec.) (lit.)

SMILES string

[Br-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Tetraethylammonium bromide (TEAB) catalyzes the synthesis of thioesters by the oxidative coupling of aldehydes or alcohols with thiols or disulfides.
  • It is used as a catalyst, along with o-iodoxybenzoic acid (IBX), in the oxidation of sulfides to sulfoxides and primary carboxamides to one-carbon dehomologated nitriles.
  • TEAB can also be used as an organic template to synthesize zeolite beta.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

361.4 °F - closed cup

Flash Point(C)

183 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetraethylammonium Bromide-Catalyzed Oxidative Thioesterification of Aldehydes and Alcohols.
Zhu, Xuebin et al.
advanced synthesis and catalysis, 355(18), 3558-3562 (2013)
Hydrothermal crystallization of zeolite beta using tetraethylammonium bromide.
Eapen M
Zeolites, 14(4), 295-302 (1994)
Masoumeh Taherimehr et al.
ChemSusChem, 10(6), 1283-1291 (2016-12-20)
Metal-organic frameworks (MOFs) with accessible Lewis acid sites are finding increasing application in the field of heterogeneous catalysis. However, the structural instability of MOFs when they are exposed to high temperature and/or high pressure often limits their applicability. In this
A mild, chemoselective oxidation of sulfides to sulfoxides using o-iodoxybenzoic acid and tetraethylammonium bromide as catalyst.
Shukla V
The Journal of Organic Chemistry, 68(13), 5422-5425 (2003)
o-Iodoxybenzoic acid-and tetraethylammonium bromide-mediated oxidative transformation of primary carboxamides to one-carbon dehomologated nitriles.
Bhalerao D
The Journal of Organic Chemistry, 72(2), 662-665 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service