Skip to Content
Merck
All Photos(2)

Key Documents

236284

Sigma-Aldrich

2-Nitroanisole

≥99%

Synonym(s):

1-Methoxy-2-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OCH3
CAS Number:
Molecular Weight:
153.14
Beilstein:
1868032
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.561 (lit.)

bp

273 °C (lit.)

mp

9-12 °C (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)
water: insoluble(lit.)

density

1.254 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

COc1ccccc1[N+]([O-])=O

InChI

1S/C7H7NO3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3

InChI key

CFBYEGUGFPZCNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Nitroanisole (2-methoxynitrobenzene is an industrial and environmental pollutant causing tumors of the urinary bladder in rats and mice. Tumorigenic potential of 2-nitroanisole has been evaluated by host-mediated in vitro and in vivo assay.

Application

2-Nitroanisole has been employed as solvatochromic probe to examine cybotactic region of pure and mixed supercritical fluid solvent systems.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markéta Miksanová et al.
Chemical research in toxicology, 17(5), 663-671 (2004-05-18)
2-Nitroanisole (2-NA) is an important industrial pollutant and a potent carcinogen for rodents. Determining the capability of humans to metabolize 2-NA and understanding which human cytochrome P450 (P450) enzymes are involved in its activation and/or detoxification are important to assess
Marie Stiborová et al.
Interdisciplinary toxicology, 2(1), 24-27 (2009-03-01)
An aromatic amine, o-anisidine (2-methoxyaniline) and its oxidative counterpart, 2-nitroanisole (2-methoxynitrobenzene), are the industrial and environmental pollutants causing tumors of the urinary bladder in rats and mice. Both carcinogens are activated to the same proximate carcinogenic metabolite, N-(2-methoxyphenyl)hydroxylamine, which spontaneously
M J Miller et al.
Drug metabolism and disposition: the biological fate of chemicals, 13(5), 527-531 (1985-09-01)
The pharmacokinetics and metabolism of o-nitroanisole (ONA) were studied in male Fischer 344 rats. Three dose levels of [14C]ONA (5.0, 50, or 500 mg/kg) were administered orally to rats and daily excreta were analyzed for 14C. Since the highest dose
J G Hengstler et al.
Scandinavian journal of work, environment & health, 21(1), 36-42 (1995-02-01)
The aim of the study was to detect single-strand breaks in deoxyribonucleic acid (DNA) in mononuclear blood cells of fire fighters exposed to o-nitroanisole and other substances released into the environment during an accident in a chemical plant. The level
Effects of physical binding of o-Nitroanisole with feed upon its systemic availability in male F344 rats.
J H Yuan et al.
Bulletin of environmental contamination and toxicology, 47(1), 152-159 (1991-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service