Skip to Content
Merck
All Photos(1)

Documents

236136

Sigma-Aldrich

Cobalt(III) fluoride

Synonym(s):

Cobalt trifluoride, Cobaltic fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoF3
CAS Number:
Molecular Weight:
115.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

concentration

48.5-52.5% Co (with A-Nitroso-B-Naphthol, gravimetric)

density

3.88 g/mL at 25 °C (lit.)

SMILES string

F[Co](F)F

InChI

1S/Co.3FH/h;3*1H/q+3;;;/p-3

InChI key

WZJQNLGQTOCWDS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service