Arrived in timely manner, correctly contained & labeled
235180
(1S,2S,5R)-(+)-Neomenthol
≥95%
Synonym(s):
(+)-Neomenthol, (1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol, D-Neomenthol, [1S-(1β,2β,5α)]-5-Methyl-2-(1-methylethyl)cyclohexanol
About This Item
Recommended Products
vapor pressure
0.8 mmHg ( 20 °C)
Quality Level
Assay
≥95%
form
liquid
optical activity
[α]22/D +17.3°, neat
refractive index
n20/D 1.461 (lit.)
bp
95 °C/12 mmHg (lit.)
mp
−22 °C (lit.)
density
0.899 g/mL at 25 °C (lit.)
functional group
hydroxyl
storage temp.
2-8°C
SMILES string
CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1O
InChI
1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10+/m1/s1
InChI key
NOOLISFMXDJSKH-UTLUCORTSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
181.4 °F - closed cup
Flash Point(C)
83 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service