Skip to Content
Merck
All Photos(2)

Documents

234788

Sigma-Aldrich

Tributyltin hydride

contains 0.05% BHT as stabilizer, 97%

Synonym(s):

TBTH, Tri-n-butylstannane, Tri-n-butyltin hydride, Tributylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnH
CAS Number:
Molecular Weight:
291.06
Beilstein:
3587329
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

0.05% BHT as stabilizer

reaction suitability

reagent type: reductant

refractive index

n20/D 1.473 (lit.)

bp

80 °C/0.4 mmHg (lit.)

density

1.082 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][Sn](CCCC)(CCCC)CCCC

InChI

1S/3C4H9.Sn.H/c3*1-3-4-2;;/h3*1,3-4H2,2H3;;

InChI key

DBGVGMSCBYYSLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A widely used radical reagent used in reductive cleavage, radical dehalogenation, and intramolecular radical cyclization.
Radical promoted intramolecular cyclization leading to isoxazolo-benzaulene ring system. Vinyl tin fragment used in a synthesis of (+)-panepophenanthrin via biomimetic Diels-Alder dimerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 9892-9892 (2006)
Synthesis, 417-417 (1988)
Synthesis, 665-665 (1987)
Tetrahedron Letters, 47, 7043-7043 (2006)
Cassander P Titley-O'Neal et al.
The Science of the total environment, 449, 52-62 (2013-02-16)
Queen conch (Strombus gigas) are listed in CITES Appendix II. Populations may be declining due to anthropogenic inputs that include pollutants from boating activity. In the British Virgin Islands (BVI), some conch exhibit imposex, a condition in which male external

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service