Skip to Content
Merck
All Photos(1)

Documents

232491

Sigma-Aldrich

5-Methoxyresorcinol

98%

Synonym(s):

3,5-Dihydroxyanisole, Phloroglucinol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
140.14
Beilstein:
1423578
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

188-189 °C/12 mmHg (lit.)

mp

78-80 °C (lit.)

SMILES string

COc1cc(O)cc(O)c1

InChI

1S/C7H8O3/c1-10-7-3-5(8)2-6(9)4-7/h2-4,8-9H,1H3

InChI key

HDVRLUFGYQYLFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of the aroxyl radical-scavenging action of 5-methoxyresorcinol has been investigated. The kinetics of reaction of 5,7-diisopropyl-tocopheroxyl radical (Toc) with 5-methoxyresorcinol has been measured by stopped-flow spectrophotometer.

Application

5-Methoxyresorcinol was used in synthesis of:
  • isorobustin
  • substituted linear and angular benzofurocoumarins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of isorobustin.
Barton DHR, et al.
Tetrahedron Letters, 31(51), 7449-7452 (1990)
Kazuo Mukai et al.
Free radical biology & medicine, 38(9), 1243-1256 (2005-04-06)
The reaction rates (k(r)) of 5,7-diisopropyl-tocopheroxyl radical (Toc) with catechins (epicatechin (EC), epicatechin gallate (ECG), epigallocatechin (EGC), epigallocatechin gallate (EGCG)) and related compounds (methyl gallate (MG), 4-methylcatechol (MC), and 5-methoxyresorcinol (MR)) have been measured by stopped-flow spectrophotometer. The k(r) values
Shuji Mitani et al.
Journal of agricultural and food chemistry, 56(12), 4406-4417 (2008-05-27)
Kinetic study of the aroxyl radical-scavenging action of catechins (epicatechin (EC), epicatechin gallate (ECG), epigallocatechin (EGC), and epigallocatechin gallate (EGCG)) and related compounds (methyl gallate (MG), 4-methylcatechol (MC), and 5-methoxyresorcinol (MR)) has been performed. The second-order rate constant ( k
Synthesis of linear and angular benzofurocoumarins.
Teran C, et al.
Synthesis, 1997(12), 1384-1386 (1997)
J U Kreft et al.
European journal of biochemistry, 226(3), 945-951 (1994-12-15)
The previously studied complete methyl transfer sequence of tetrahydrofolate-dependent O-demethylation catalyzed by Holophaga foetida strain TMBS4 extracts was separated into two steps using cobalamins as non-physiological substrates: electrochemically produced cob(I) alamin served as methyl acceptor for phenyl methyl ether demethylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service