232041
Cesium hydroxide solution
50 wt. % in H2O, 99.9% trace metals basis
Synonym(s):
Caesium hydroxide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CsOH
CAS Number:
Molecular Weight:
149.91
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
liquid
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
liquid
concentration
50 wt. % in H2O
impurities
≤1000.0 ppm Trace Metal Analysis
density
1.72 g/mL at 25 °C
SMILES string
[OH-].[Cs+]
InChI
1S/Cs.H2O/h;1H2/q+1;/p-1
InChI key
HUCVOHYBFXVBRW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Cesium hydroxide solution is used to make porous cesium titanosilicates, potential ion-exchange materials for cleanup of radioactive wastes.CsOH impregnated in FAU zeolites enhanced their basic properties without affecting the FAU framework.It can effectively catalyse the condensation of methyl propionate with formaldehyde to yield methyl methacrylate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Formation of methyl methacrylate by condensation of methyl propionate with formaldehyde over silica-supported cesium hydroxide catalysts
Ai M
Applied Catalysis A: General, 288(1-2), 211-215 (2005)
Behrens, E.A. et al.
Chemistry of Materials, 8, 1236-1236 (1996)
Enhancement of the basic properties in FAU zeolites by impregnation with cesium hydroxide
Romero MD, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 81(1-3), 313-320 (2005)
Felix Neumaier et al.
The Journal of general physiology, 152(9) (2020-06-20)
Loosely bound Zn2+ ions are increasingly recognized as potential modulators of synaptic plasticity and neuronal excitability under normal and pathophysiological conditions. Cav2.3 voltage-gated Ca2+ channels are among the most sensitive targets of Zn2+ and are therefore likely to be involved
M M V Ramana et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 165-171 (2015-07-25)
In the present work, isopropyl-6-amino-4-(3,5-bis(trifluoromethyl)phenyl)-5-cyano-2-methyl-4H-pyran-3-carboxylate (4H-pyran analog) has been synthesized by a three component reaction catalyzed by CsOH/γ-Al2O3 and characterized. The interaction of 4H-pyran analog with herring sperm DNA (hs DNA) under physiological conditions (phosphate buffer of pH 7.2) was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service