Skip to Content
Merck
All Photos(2)

Key Documents

229857

Sigma-Aldrich

Selenous acid

99.999% trace metals basis

Synonym(s):

Selenious acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SeO3
CAS Number:
Molecular Weight:
128.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

mp

70 °C (dec.) (lit.)

density

3.004 g/mL at 25 °C (lit.)

SMILES string

O[Se](O)=O

InChI

1S/H2O3Se/c1-4(2)3/h(H2,1,2,3)

InChI key

MCAHWIHFGHIESP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Selenous acid is a dibasic acid. It is the oxoacid of selenium and can be formed by adding selenium dioxide gas to water. Selenous acid shows analogy to sulfurous acid. As a crystalline solid, the molecules are arranged in a pyramid and in a solution it behaves as a diprotic acid.

Application

  • Selenic acid etching assisted vacancy engineering for designing highly active electrocatalysts toward the oxygen evolution reaction: Examination of the use of selenous acid in vacancy engineering for electrocatalysts (Zhang et al., 2021).
  • Europium (III) Catalysis for Reduction of Thionine Dye by Selenous Acid in Aqueous Sulfuric Acid Solutions: Study on the kinetics and mechanism of selenium(IV) reduction in aqueous sulfuric acid solutions (Fawzy, 2016).

Packaging

Packaged in High-Density Polyethylene (HDPE)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ropp RC et al.
Encyclopedia of the Alkaline Earth Compounds null
Thomas G Back et al.
Journal of the American Chemical Society, 124(41), 12104-12105 (2002-10-10)
1,2-Oxaselenolane Se-oxide is a novel cyclic seleninate ester that functions as a remarkably efficient glutathione peroxidase mimetic by catalyzing the reduction of tert-butyl hydroperoxide to tert-butyl alcohol in the presence of benzyl thiol. The seleninate ester can be conveniently generated
Thomas G Back et al.
The Journal of organic chemistry, 70(23), 9230-9236 (2005-11-05)
[Reaction: see text]. Several novel organoselenium and tellurium compounds were prepared and evaluated as mimetics of the selenoenzyme glutathione peroxidase, which protects cells from oxidative stress by reducing harmful peroxides with the thiol glutathione. The compounds were tested for catalytic
H Chen et al.
International journal of food microbiology, 18(2), 151-159 (1993-04-01)
Five Salmonella serotypes recovering from heat injury exhibited different kinetics of resuscitation and growth. Exponential growth was reached before full resuscitation. Fully resuscitated cells and uninjured cells at low cell densities exhibited sensitivity when transferred from non-selective media to selective
M Rizki et al.
Environmental and molecular mutagenesis, 37(1), 70-75 (2001-02-15)
The genotoxic activity of three selenium compounds (sodium selenite, sodium selenate, and selenious acid) and the antigenotoxic effects of sodium selenite in combination with the chromium compound potassium dichromate were studied using the wing spot test of Drosophila melanogaster. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service