The particle size specification for this material is simply <10 μm. The particle size distribution has not been determined.
221643
Cobalt(II,III) oxide
powder, <10 μm
Synonym(s):
Cobaltosic oxide, Tricobalt tetraoxide
About This Item
Recommended Products
form
powder
Quality Level
particle size
<10 μm
mp
895 °C (dec.) (lit.)
density
6.11 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
O=[Co].O=[Co]O[Co]=O
InChI
1S/3Co.4O
InChI key
LBFUKZWYPLNNJC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Synthesis of diphenylalanine/cobalt oxide hybrid nanowires and their application to energy storage.: This study demonstrates the synthesis of hybrid nanowires using diphenylalanine and cobalt oxide, highlighting their potential for enhancing energy storage capacities (Ryu et al., 2010).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Carc. 1A Inhalation - Resp. Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Do you have the particle size distribution graph for this product?
1 answer-
Helpful?
-
-
Where is the purity?
1 answer-
Unfortunately, the purity is not a test specification for this product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service