Skip to Content
Merck
All Photos(1)

Key Documents

218979

Sigma-Aldrich

4-(Dimethylamino)cinnamic acid

99%

Synonym(s):

3-(4-Dimethylaminophenyl)acrylic acid, 3-[4-(Dimethylamino)phenyl]-2-propenoic acid, 4-(Dimethylamino)cinnamic acid, p-(Dimethylamino)cinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
191.23
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

powder

mp

227-228 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CN(C)c1ccc(\C=C\C(O)=O)cc1

InChI

1S/C11H13NO2/c1-12(2)10-6-3-9(4-7-10)5-8-11(13)14/h3-8H,1-2H3,(H,13,14)/b8-5+

InChI key

CQNPVMCASGWEHM-VMPITWQZSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noah D Bronstein et al.
ACS nano, 13(4), 3839-3846 (2019-03-12)
We present a combined experimental and theoretical study of ligand-ligand cooperativity during X-type carboxylate-to-carboxylate ligand exchange reactions on PbS quantum dot surfaces. We find that the ligand dipole moment (varied through changing the substituents on the benzene ring of cinnamic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service