21861
1,1′-Carbonyl-di-(1,2,4-triazole)
technical, ≥90% (T)
Synonym(s):
CDT
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H4N6O
CAS Number:
Molecular Weight:
164.12
Beilstein:
10250
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Quality Level
Assay
≥90% (T)
form
powder
reaction suitability
reaction type: Carbonylations
mp
138-152 °C
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
O=C(n1cncn1)n2cncn2
InChI
1S/C5H4N6O/c12-5(10-3-6-1-8-10)11-4-7-2-9-11/h1-4H
InChI key
YHNUDLCUIKMNSN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Reagent for preparing the reactive acyl triazolides from carboxylic acids; Reaction of unprotected glucose with acyl triazolide to the β-D-glucosyl ester; Dehydrating agent which reacts with H2O2 giving singlet oxygen, used e.g. for epoxidations
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Samantha B Peeters et al.
Human molecular genetics, 27(7), 1252-1262 (2018-02-06)
A long-standing question concerning X-chromosome inactivation (XCI) has been how some genes avoid the otherwise stable chromosome-wide heterochromatinization of the inactive X chromosome. As 20% or more of human X-linked genes escape from inactivation, such genes are an important contributor
J. Rebek et al.
The Journal of Organic Chemistry, 44, 1485-1485 (1979)
Yan Li et al.
Pharmaceutical biology, 54(12), 3226-3231 (2016-10-01)
A classic traditional Chinese medicine Curcuma aromatica Salisb. (Zingiberaceae) has been reported to have favourable effects on the cardiovascular system. To research the cardio-protective effect of different C. aromatica hydroalcoholic extracts on isoproterenol (ISO)-induced acute myocardial ischemia (AMI) in rats.
Steen Buus Kristiansen et al.
Cardiovascular diabetology, 18(1), 66-66 (2019-06-04)
The mechanisms underlying increased mortality in patients with diabetes and admission hyperglycemia after an acute coronary syndrome may involve reduced capacity for cardioprotection. We investigated the impact of hyperglycemia on exogenously activated cardioprotection by ischemic preconditioning (IPC) in hearts from
Larissa Haertle et al.
PloS one, 12(8), e0184030-e0184030 (2017-08-31)
Imprinted genes show parent-specific activity (functional haploidy), which makes them particularly vulnerable to epigenetic dysregulation. Here we studied the methylation profiles of oppositely imprinted genes at single DNA molecule resolution by two independent parental allele-specific deep bisulfite sequencing (DBS) techniques.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service