Skip to Content
Merck
All Photos(1)

Key Documents

217727

Sigma-Aldrich

4-Chloro-2-(trifluoromethyl)aniline

97%

Synonym(s):

2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)NH2
CAS Number:
Molecular Weight:
195.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.507 (lit.)

bp

66-67 °C/3 mmHg (lit.)

density

1.386 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(Cl)cc1C(F)(F)F

InChI

1S/C7H5ClF3N/c8-4-1-2-6(12)5(3-4)7(9,10)11/h1-3H,12H2

InChI key

CVINWVPRKDIGLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fourier-transform (FT) infrared and FT-Raman spectra of 4-chloro-2-(trifluoromethyl) aniline has been studied.

Application

4-Chloro-2-(trifluoromethyl)aniline was used in the synthesis of 2-amino-5-chloro 3-(trifluoromethyl)benzenethiol.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-amino-5-chloro-3-(trifluoromethyl) benzenethiol and conversion into 4H-1, 4-benzothiazines and their sulfones.
Thomas L, et al.
Journal of Fluorine Chemistry, 122(2), 207-213 (2003)
M Arivazhagan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 205-213 (2011-11-15)
The Fourier-transform infrared and FT-Raman spectra of 4-chloro-2-(trifluoromethyl) aniline (4C2TFA) were recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1) respectively. Quantum chemical calculations of energies, geometrical structure and vibrational wavenumbers of 4C2TFA were carried out by density functional theory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service