Skip to Content
Merck
All Photos(2)

Key Documents

215414

Sigma-Aldrich

Calcium

turnings, 99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca
CAS Number:
Molecular Weight:
40.08
Beilstein:
4241647
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis

form

turnings

reaction suitability

reagent type: reductant

resistivity

3.5 μΩ-cm, 20°C

bp

1484 °C (lit.)

mp

850 °C (lit.)

density

1.54 g/mL at 25 °C (lit.)

SMILES string

[Ca]

InChI

1S/Ca

InChI key

OYPRJOBELJOOCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsai-Wen Chen et al.
Nature, 499(7458), 295-300 (2013-07-23)
Fluorescent calcium sensors are widely used to image neural activity. Using structure-based mutagenesis and neuron-based screening, we developed a family of ultrasensitive protein calcium sensors (GCaMP6) that outperformed other sensors in cultured neurons and in zebrafish, flies and mice in
S Boonen et al.
Journal of internal medicine, 259(6), 539-552 (2006-05-18)
Combined calcium and vitamin D supplementation is an essential component of the management of osteoporosis, supported by a strong scientific rationale. The types of individuals who should receive calcium and vitamin D supplements are those: (i) patients with documented osteoporosis
Byrappa Venkatesh et al.
Nature, 505(7482), 174-179 (2014-01-10)
The emergence of jawed vertebrates (gnathostomes) from jawless vertebrates was accompanied by major morphological and physiological innovations, such as hinged jaws, paired fins and immunoglobulin-based adaptive immunity. Gnathostomes subsequently diverged into two groups, the cartilaginous fishes and the bony vertebrates.
Eleni Gourni et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(10), 1719-1725 (2014-08-26)
Gastrin-releasing peptide receptors (GRPrs) are overexpressed on a variety of human cancers, providing the opportunity for peptide receptor targeting via radiolabeled bombesin-based peptides. As part of our ongoing investigations into the development of improved GRPr antagonists, this study aimed at
Marco Antunes et al.
The Journal of cell biology, 202(2), 365-379 (2013-07-24)
Epithelial wound healing relies on tissue movements and cell shape changes. Our work shows that, immediately after wounding, there was a dramatic cytoskeleton remodeling consisting of a pulse of actomyosin filaments that assembled in cells around the wound edge and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service