About This Item
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.585 (lit.)
bp
93-95 °C/0.2 mmHg (lit.)
density
1.139 g/mL at 25 °C (lit.)
functional group
sulfoxide
storage temp.
2-8°C
SMILES string
C=CS(=O)c1ccccc1
InChI
1S/C8H8OS/c1-2-10(9)8-6-4-3-5-7-8/h2-7H,1H2
InChI key
MZMJHXFYLRTLQX-UHFFFAOYSA-N
Gene Information
human ... LOC129293(129293)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service