Recommended Products
form
powder
Quality Level
composition
Dye content, 50%
λmax
594 nm
ε (extinction coefficient)
≥15000 at 283-289 nm in water at 0.01 g/L
≥30000 at 582-588 nm in water at 0.01 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Na+].[Na+].[Na+].[Na+].Nc1ccc2c(O)c(N=Nc3ccc(N=Nc4ccc(N=Nc5cc(c6cccc(c6c5)S([O-])(=O)=O)S([O-])(=O)=O)c7ccccc47)c8ccc(cc38)S([O-])(=O)=O)c(cc2c1)S([O-])(=O)=O
InChI
1S/C40H27N7O13S4.4Na/c41-22-8-10-25-21(16-22)17-38(64(58,59)60)39(40(25)48)47-46-35-15-14-34(28-11-9-24(20-30(28)35)61(49,50)51)45-44-33-13-12-32(26-4-1-2-5-27(26)33)43-42-23-18-31-29(37(19-23)63(55,56)57)6-3-7-36(31)62(52,53)54;;;;/h1-20,48H,41H2,(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60);;;;/q;4*+1/p-4
InChI key
QTTDXDAWQMDLOF-UHFFFAOYSA-J
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service