205478
Bromoxylenol Blue
indicator grade, Dye content 95 %
Synonym(s):
3′,3″-Dibromo-p-xylenolsulfonphthalein
About This Item
Recommended Products
grade
indicator grade
form
powder or crystals
composition
Dye content, 95%
technique(s)
titration: suitable
pH
6.0-7.6, yellow to blue
mp
218 °C (dec.) (lit.)
solubility
ethanol: 1 mg/mL, clear
λmax
417 nm
ε (extinction coefficient)
≥16000 at 417-421 nm in methanol at 0.016 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Cc1cc(c(C)c(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(Br)c4C
InChI
1S/C23H20Br2O5S/c1-11-9-16(13(3)19(24)21(11)26)23(17-10-12(2)22(27)20(25)14(17)4)15-7-5-6-8-18(15)31(28,29)30-23/h5-10,26-27H,1-4H3
InChI key
MRDOFVRMTNWMDA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recent applications of bromoxylenol blue include:
- Building pH sensors
- Developing optical sensors for high acidity measurements
- Synthesizing antifungal activity compounds
- Determination of drugs
- In fiber-optic biosensors for the determination and detection of pesticides.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service